Tisk…
Measuring beakers with handle
Dodavatel: RSG
077.001 077.002 077.003 077.005 077.500 077.250 077.100
213-1037EA 12900 CZK
213-1037 213-1038 213-1039 213-1040 213-0376 213-0735 213-0374
Measuring beakers with handle
Kádinky
Stainless steel, rust-proof, with flared rim, spout and handle.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.