Tisk…
Sample tubes
Dodavatel: Simport Scientific
T500-2T T500-4AT T500-4T T500-5T T500-1T
212-9331EA 3910 CZK
212-9331 212-9332 212-9333 212-9334 212-9324
Sample tubes
Zkumavky Zkumavky na vzorky
PP, translucent
  • Withstand temperatures from −196 to 121 °C
  • Withstand centrifugation
  • Autoclavable
  • Non sterile
For storing and transporting biological material. Tube with internal threads. Caps are supplied with or without a silicone washer, to ensure a positive leakproof seal at all temperatures. A 1 1/4 turn of the cap is sufficient to seal the vial.
Since both closures and tubes are manufactured of the same material, they have the same coefficient of expansion to guarantee an equally secure seal both at room or at low temperatures. Round bottomMore Product Information