Tisk…
Beakers, triangular, Tricorn™
Dodavatel: Simport Scientific
B700-100 B700-1L B700-250 B700-400 B700-800 B700-50
212-9302EA 1450 CZK
212-9302 212-9306 212-9303 212-9304 212-9305 212-9301
Beakers, triangular, Tricorn™
Kádinky
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Balení: All sizes are available 100 per case.
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904