Tisk…
VWR® Reduction Adapters for Conical Ground Joints
Dodavatel: VWR Collection
212-0858EA 309 CZK
212-0858 212-0859 212-0860 212-0861 212-0862 212-0863 212-0864 201-1436 201-1437 201-1438
VWR® Reduction Adapters for Conical Ground Joints
Adaptéry Adaptéry s kónickým zábrusů
Adapters from large cone at the top to small socket at the bottom.
  • Made from clear borosilicate 3.3 glass
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Outstanding leak tightness
Certifikace: Produced according to DIN 12257.