Tisk…
HFC39 series plastic couplings
Dodavatel: CPC COLDER
HFC22439M HFC22839M HFCD17439M HFCD17639M HFCD17839M HFCD22439M HFCD22639M HFCD22839M
212-0793EA 7190 CZK
212-0793 212-0794 212-0795 212-0796 212-0797 212-0798 212-0799 212-0800
HFC39 series plastic couplings
Fitinky a konektory
These high flow couplings feature automatic shut-off valves which stop flow and eliminate the need for pinch clamps, stainless steel connectors or fittings.
They allow rapid disconnect and reconnect without the need for tools. Valved couplings stop fluid flow at the point of connection, reducing spills and related safety issues. Couplings feature an audible click when engaged, ensuring the user of a secure connection. Constructed of medical grade polysulphone (amber tint) with clear silicone seals. Couplings are sterilisable, reusable and compatible with virtually all existing processes.
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Maximální provozní tlak Vacuum...8,62 bar
Provozní teplota –40...+137,8 °C