Tisk…
VWR®, Test Tubes with Rim
Dodavatel: VWR Collection
212-0753EA 809 CZK
212-0753 212-0754 212-0755 212-0756 212-0757 212-0758 212-0759 212-0760 212-0761 212-0762 212-0763 212-0764 212-0765
VWR®, Test Tubes with Rim
Zkumavky Zkumavky na vzorky
Test tubes made from borosilicate 5.1 glass, neutral glass.
  • Thin walled
  • Excellent chemical resistance
  • Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses.
Certifikace: DIN 12395.