Tisk…
Plugs for test tubes
Dodavatel: BOETTGER PAUL
01.604.0102 01.604.0100 01.624.0100 01.624.0101 01.624.0102 01.624.0103 01.624.0104 01.604.0101 01.604.0103 01.604.0104
212-0520EA 852 CZK
212-0520 212-0518 212-0523 212-0524 212-0525 212-0526 212-0527 212-0519 212-0521 212-0522
Plugs for test tubes
Uzávěry Víčka na zkumavky
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
  • Not autoclavable
  • Easy handling with a high, ribbed grip
  • Easy to open by tilting 'one-hand operation'