Tisk…
Magnetic Beads, Sera-Mag SpeedBeads Streptavidin-blocked magnetic particles
Dodavatel: Cytiva
21152104010150 21152104010350 21152104011150
21152104010150.EA 28310 CZK
21152104010150. 21152104010350. 21152104011150.
Magnetic Beads, Sera-Mag SpeedBeads Streptavidin-blocked magnetic particles
Kuličky Kuličky magnetické
Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine high affinity for biotinylated target molecules with very low non specific binding. Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine fast reaction kinetics with very low non specific binding for increased throughput and precision. Applications include automated immunoassays, immunoprecipitation, and protein purification. Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation. Sera-Mag SpeedBeads and Sera-Mag Streptavidin-Coated Magnetic ParticlesMore Product Information
Objednat
Produktová specifikace
Kapacita vázání 3000 - 4000 pmole/mg beads
Environmentally Preferable
Velikost částic nominal 1 µm diameter