Tisk…
PCR microtube strips, 0,2 ml, Amplitube™
Dodavatel: Simport Scientific
T320-1B T320-1G T320-1N T320-1R T320-1Y T321-1N T321-1B T321-1G T321-1R T321-1Y T321-2N T320-10N T321-2B T321-2G T321-2R T321-2Y T321-3N
211-3322EA 3230 CZK
211-3322 211-3323 211-3321 211-3324 211-3325 211-3326 211-3327 211-3328 211-3329 211-3330 SIMPT321-2N SIMPT320-10N SIMPT321-2B SIMPT321-2G SIMPT321-2R SIMPT321-2Y SIMPT321-3N
PCR microtube strips, 0,2 ml, Amplitube™
Zkumavky Zkumavky pro PCR
PP. Ultra-thin wall design ensures uniform and efficient thermal transfer, therefore reducing PCR reaction time in most V-bottomed thermal cyclers.
  • Will fit standard well spacing and can be used with 8-channel pipettes
  • Strips can be cut and used as individual tubes
  • Choice of natural and four non-cytotoxic and non-metallic colours for visual coding of samples
  • Non-attached, domed cap strips, which must be ordered separately, ensure closure during the whole thermal cycle
  • Certified RNase, DNase and DNA-free
Ultra clear, flat caps are ideal forMore Product Information