Tisk…

Oxid hlinitý, Durmax® for metallography, 15 minutes

Dodavatel: ALUMINES DURMAX

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

20987.265
20987.265EA 0 CZK
20987.265
Oxid hlinitý, Durmax® for metallography, 15 minutes
Oxid hlinitý

Informace o dodávce: This product is not available in all countries. Please check with your local VWR International office or supplier.

Vzorec: Al₂O₃
Bod varu: 2980 °C (1013 hPa)
Bod tání: 2045 °C
Hustota: 3,94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR