Tisk…
Anti- Llama Polyclonal Antibody (CF®583R)
Dodavatel: Biotium
20909-500UL 20909-50UL
20909-500ULEA 6200 CZK
20909-500UL 20909-50UL
Anti- Llama Polyclonal Antibody (CF®583R)
Protilátky
Llama anti-rabbit IgG (H L) secondary antibody labeled with our superior CF® Dyes.
  • Available in 6 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • HRP or DNP conjugates: add 1 ml distilled water
  • CF® Dye and biotin conjugates: add 0.5 ml distilled water
Add the indicated volume of water directly to the vial containing the lyophilized antibody and mix gently to dissolve. Store reconstituted antibody at -20°C and protect from light. Aliquot to avoid repeated freeze/thaw cycles. Alternatively,More Product Information