Tisk…
Anti-IgG Goat Polyclonal Antibody (CF® 583R)
Dodavatel: Biotium
20905-500UL 20905-50UL
20905-500ULEA 7050 CZK
20905-500UL 20905-50UL
Anti-IgG Goat Polyclonal Antibody (CF® 583R)
Protilátky
This is a highly cross-adsorbed goat anti-rabbit IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes. To minimise cross-reactivity, the antibody has been adsorbed against human, mouse, and rat serum.
  • Highly cross-adsorbed for specific staining with minimal background
  • Available in 19 bright and photostable CF® Dyes
  • Biotin and HRP conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
Liquid format: Store at −20 °C, protected from light. Product is stable for at least 6 monthsMore Product Information
Objednat
Produktová specifikace
Symbol antigenu IgG
Environmentally Preferable
Druh protilátky Sekundární
Klonovalita Polyclonal
Spojení CF® 583R
Reaktivita Rabbit
Hostitel Goat
Izotyp IgG
Western blot Yes
ELISA Yes
ImunoFluorescence Yes
Průtoková cytometrie Yes
Koncentrace 2 mg/ml, 1 mg/ml (HRP, AP conjugates)