Tisk…

2-Propanol 99,0, GPR RECTAPUR®

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

20839.297EA 910 CZK
20839.297 20839.366 20839.468 20839.550 20839.930
2-Propanol 99,0, GPR RECTAPUR®
2-Propanol
Vzorec: (CH₃)₂CHOH
MW: 60.1
Bod varu: 82
Bod tání: –89 °C
Hustota: 0.786 g/cm³ (20 °C)
Bod vzplanutí: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS číslo: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay Min. 99.0 %
Acidity Passes test
Acidity or alkalinity Passes test
Benzene and related substances Passes test
IR Spectrum Passes test
Peroxides Passes test
Boiling point 82.2 - 82.6 °C
Density (20/20) 0.785 - 0.789
Density (25/25) 0.783 - 0.790
Refractive index (20°C) 1.376 - 1.378
Evaporation residue Max. 20 ppm
Water Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR