Tisk…

jod ≥99.8%, solid ACS, Fluka™

Dodavatel: Honeywell Chemicals
Nebezpečí

207772-100G 207772-2.5KG 207772-500G 207772-1KG
207772-100GEA 3360 CZK
207772-100G 207772-2.5KG 207772-500G 207772-1KG
jod ≥99.8%, solid ACS, Fluka™
jod

Honeywell ACS solvents for general laboratory use are:


  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested meet ACS general use specifications

Vzorec: I₂
Hustota: 4,930 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS číslo: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III

Objednat

Výsledky specifikačního testu

Assay Min. 99.8 %
Non-volatile matter Max. 0.01 %
Bromine and chlorine (as chloride) Max. 0.005 %
Form Solid
Vapor Pressure 0.31 mmHg ( 25 °C)
Resistivity 1.3E15 μΩ-cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR