Tisk…

EDTA didraselná sůl dihydrát 97,0 zA

Dodavatel: VWR Chemicals

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt

20490.188EA 1690 CZK
20490.188
EDTA didraselná sůl dihydrát 97,0 zA
EDTA didraselná sůl dihydrát
Vzorec: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46
Bod tání: 272
Hustota: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS číslo: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546

Objednat

Výsledky specifikačního testu

Assay Min. 97.0 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 0.05 %
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR