Tisk…
Anti-Myc Tag Rabbit Polyclonal Antibody (CF594)
Dodavatel: Biotium
20438
20438.EA 9260 CZK
20438.
Anti-Myc Tag Rabbit Polyclonal Antibody (CF594)
Protilátky
Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™594 dye. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields muchMore Product Information