Tisk…
Anti-IgY Goat Polyclonal Antibody (CF™ 405M)
Dodavatel: Biotium
20375-50UL
20375-50ULEA 4280 CZK
20375-50UL
Anti-IgY Goat Polyclonal Antibody (CF™ 405M)
Protilátky
CF™405 antibodies are affinity-purified antibodies labeled with blue fluorescent CF™405S or CF™405M, two members of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™405S and CF™405M are two blue fluorescent dyes optimized for the 405nm laser line. CF™405S has excitation/emission maxima at 404/431nm and is spectrally similar toMore Product Information