Tisk…
Bottle heads, adjustable for gas washing bottles, Drechsel pattern, Quickfit®
Dodavatel: SciLabware
MF27/3/13
201-8629EA 1620 CZK
201-8629
Bottle heads, adjustable for gas washing bottles, Drechsel pattern, Quickfit®
Uzávěry Uzávěry k láhvím
PYREX®, borosilicate glass, without glass filter disc.
  • Very good chemical resistance
  • High temperature resistance
With screwthread joint which allows height adjustment of the dip tube and hence permits use with either 125 ml or 250 ml Drechsel bottles. Detachable plastic screwthreads connectors for easy fitting of flexible tubing.