Tisk…
Flat flange lids with 5 necks
Dodavatel: Lenz Laborglas GmbH & CO.KG
06105244 06105464 06105444 06105241 06105264 06105544 06105564
LENZ06105244EA 8710 CZK
LENZ06105244 LENZ06105464 201-1143 201-1142 LENZ06105264 LENZ06105544 LENZ06105564
Flat flange lids with 5 necks
Víčka Reaction Vessel Lids
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat laboratory flange (DN). Straight middle neck, three angled side necks and one straight side neck with standard ground joints.