Tisk…
Adapters, distilling heads
Dodavatel: Lenz Laborglas GmbH & CO.KG
05028011 05028022 05028032 05028033 05028044
201-0338EA 763 CZK
201-0338 201-0339 201-0340 201-0341 201-2222
Adapters, distilling heads
Komponenty pro destilaci Hlavice
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Includes a socket for a distillation thermometer of 55 mm fitted length and two cones at an angle of 75°.