Tisk…
O-rings for flat flanges, DURAN®
Dodavatel: DWK Life Sciences
292223406 292224608 292226103 292225107 292225707
273-0161EA 1490 CZK
273-0161 201-0162 273-0165 201-0163 201-0164
O-rings for flat flanges, DURAN®
Těsnění Těsnící kroužky
Silicone core sheathed with FEP, red.
  • Guarantee excellent joint seal
  • Temperature resistant from −200 to +200 °C
  • Excellent chemical resistance
O-rings for flat flanges with groove.