Tisk…
Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2.43]
Dodavatel: Tonbo Biosciences
20-1886-U100 20-1886-U025
20-1886-U100EA 5020 CZK
20-1886-U100 20-1886-U025
Anti-CD8A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2.43]
Protilátky
The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils.
In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes. Note: forMore Product Information
Objednat
Produktová specifikace
Symbol antigenu CD8A
Environmentally Preferable
Název antigenu CD8 antigen, alpha chain
Synonyma antigenu p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
Druh protilátky Primární
Klonovalita Monoclonal
Spojení APC (Allophycocyanin)
Klon 2.43
Reaktivita Mouse
Hostitel Rat
Identifikační číslo genu 12525
Izotyp IgG2b
Imunochemie Yes
Průtoková cytometrie Yes
Křížová adsorbce none
Skladovací teplota Cool
Transportní teplota Ice