Tisk…
Dulbecco's phosphate-buffered saline (DPBS), Corning®
Dodavatel: Corning
55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
55-031-PBEA 1080 CZK
55-031-PB 55-031-PC 20-030-CV 20-031-CV 21-030-CV 21-031-LX 21-031-LB 21-031-CV 21-031-CM 21-030-CM
Dulbecco's phosphate-buffered saline (DPBS), Corning®
Pufry Pufry pro tkáňové kultury
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7,0 to 7,6
  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Certifikace: Complete Certificate of Analysis available for each production lot, along with full formulation information.
Upozornení: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
Objednat
Produktová specifikace
Environmentally Preferable
Obsah endotoxinů