Tisk…
Disk filters, Whatman™ VACU-GUARD
Dodavatel: Whatman products (Cytiva)
6722-5000 6722-5001
189-0303EA 6830 CZK
189-0303 189-0304
Disk filters, Whatman™ VACU-GUARD
Filtry Filtry In-Line
Whatman VACU-GUARD disk filters are ideal for protecting vacuum pump systems from aqueous aerosols and water intrusion.
  • Designed for in-line use with stepped barb connections
  • Membrane retains 99,99% of airbone particles greater than 0,1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane
Available in 50 and 60 mm formats.
Informace pro objednávání: All materials pass USP Class VI test for plastics.