Tisk…
Filtry na ochranu pump a aparatur, VACU-GUARD™ a VACU-GUARD™ 150, Whatman™
Dodavatel: Whatman products (Cytiva)
6722-1001 6722-1002
189-0300EA 6380 CZK
189-0300 189-0301
Filtry na ochranu pump a aparatur, VACU-GUARD™ a VACU-GUARD™ 150, Whatman™
Filtry Filtry In-Line
Whatman VACU-GUARD capsule filters are ideal for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
  • Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols
  • Designed for in-line use with stepped barb connections for 10 to 12 mm ID hose
  • Available with choice of chemical trap: Activated carbon, molecular sieve, or desiccant
  • Membrane retains 99,99% of airbone particles greater than 0,1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane