Tisk…

Lutetium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific

18685.03 18685.06
18685.03EA 5080 CZK
18685.03 18685.06
Lutetium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lutetium trichloride

Purity: 99.9% CAS No.: 10099-66-8 Molecular Formula: LuCl3 Formula Weight: 281.33 Physical Form: -20 Mesh Density: 3.98 Melting Point: 905° subl. Merck Reference: 14,5615 MDL No.: MFCD00011099

Vzorec: Cl₃Lu
Bod tání: 905 °C (subl.)
Hustota: 3,98
Storage Temperature: Ambient
MDL Number: MFCD00011099
CAS číslo: 10099-66-8
EINECS: 233-240-1
Merck Index: 14,5615

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR