Tisk…

Petrolether, 35…60 °C ACS, Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

184519-1L 184519-6X1L 184519-2.5L
184519-1LEA 2470 CZK
184519-1L 184519-6X1L 184519-2.5L
Petrolether, 35…60 °C ACS, Riedel-de Haën™
Ropný líh

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


UNSPSC Code: 12191502

Bod varu: 40…60 °C (1.013 hPa)
Hustota: 0.65 g/cm³ (20 °C)
Bod vzplanutí: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS číslo: 64742-49-0
EINECS: 265-151-9
UN: 3295
ADR: 3 II

Objednat

Výsledky specifikačního testu

Identity (IR) complying
Refractive index (n 20/D) 1.358 - 1.368
Non-volatile matter Max. 0.001 %
Boiling range 35 - 60 °C
Explosion Limit 8 %
Acidity complying
APHA Max. 10
Autoignition Temperature 475 °F
Vapor Pressure 7.99 psi ( 20 °C)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR