Tisk…
Diaphragm pumps
Dodavatel: VACUUBRAND
20696080 696082 20738000 20731000 20731100 20732000 20732200 20734000 20736000 20721000 20730000 20741000 20743000 20744000
181-1580EA 53010 CZK
181-1580 181-0016 181-1082 181-1033 181-1034 181-1037 181-1039 181-1061 181-1065 181-0241 181-0476 181-0477 181-0478 181-0437
Diaphragm pumps
Čerpadla Pumpy vakuové membránové
These mechanical diaphragm pumps function without oil. They are easy to operate, use no water and generate neither wastewater nor used oil. These pumps are used for evacuating or pumping gases between containers in chemistry or physics laboratories and are extremely versatile.
  • Available in aluminium or chemical
  • Large selection of final pressure and pump capacities, high pump volume capacities even at low pressures
  • Maintenance-free drive unit, constructed for easy maintenance, diaphragm and valves are easy to replace
  • Very quiet, low-vibration
  • Optimised
More Product Information