Tisk…
e.jet pump, Microsart®
Dodavatel: Sartorius
166MP-4
181-0589EA 48450 CZK
181-0589
e.jet pump, Microsart®
Čerpadla Pumpy vakuové
The Microsart® e.jet is a laboratory vacuum pump which creates vacuum and concomitantly transfers the filtered liquid to waste.
  • For vacuum filtration in microbiological analysis
Microsart® e.jet is ideal for sample preparation in microbiological analysis achieving a trans membrane pressure of 600 mbar and a flow rate of 4,0 Nl/min (4,0 Normlitre water displacement by air in 1 minute). Constant flow rates and a defined maximum vacuum guarantees smooth and reliable filtration.