Tisk…
Vacuum Controller, V-850/V-855
Dodavatel: BUCHI
047231 047160 047299
531-0154EA 42610 CZK
531-0154 181-0140 531-0232
Vacuum Controller, V-850/V-855
Čerpadla Kontroléry vakua
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option ofMore Product Information