Tisk…
Diaphragm pumps, VARIO-SP®
Dodavatel: VACUUBRAND
20696101 20696110
181-0017EA 57910 CZK
181-0017 VACU20696110
Diaphragm pumps, VARIO-SP®
Čerpadla Pumpy vakuové membránové
The three-stage VARIO-SP® diaphragm pumps, with its variable speed drive system, attains significantly higher pumping speed and a better ultimate vacuum than the corresponding fixed speed pump. The brushless motor (maintenance-free) enables easy integration of the pump into other equipment, and operation independent of line voltage. The motor speed is either fixed or can be adjusted externally via electronic control signals for adaptation to process requirements.
  • Contamination-free pumping and evacuation
  • High flow rate even near ultimate vacuum
  • Fast pump
More Product Information