Tisk…
VWR®, Water Purification System, E Series
Dodavatel: VWR Collection
171-1251EA 277820 CZK
171-1251 171-1252 171-1253 171-1473 171-1474 171-1475
VWR®, Water Purification System, E Series
Systémy pro výrobu čisté vody Systémy pro výrobu čisté a ultračisté vody
This compact system produces EDI-based Type II water directly from tap water. Quality of Type II water meets or exceeds water quality as defined by ASTM, CAP, CLSI and ISO 3696 / BS 3997, and also complies with the purified water requirements from the European and U.S. Pharmacopoeia.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow
More Product Information
Objednat
Produktová specifikace
Model E5 E10 E15
Environmentally Preferable
Přiváděná voda Tap water (conductivity: <2000 µS/cm / TDS: <1000 ppm) Tap water (conductivity: <2000 µS/cm / TDS: <1000 ppm) Tap water (conductivity: <2000 µS/cm / TDS: <1000 ppm)
Průtok 5 L/h,Max. 2 l/min 10 L/h,Max. 2 l/min 15 L/h,Max. 2 l/min
Průtok 5 L/h,Max. 2 l/min 10 L/h,Max. 2 l/min 15 L/h,Max. 2 l/min
TOC content <30 ppb <30 ppb <30 ppb
Resistivity at +25 °C >5 MΩ·cm (typically 10 - 15 MΩ·cm) >5 MΩ·cm (typically 10 - 15 MΩ·cm) >5 MΩ·cm (typically 10 - 15 MΩ·cm)
Š×H×V 320×440×540 mm 320×440×540 mm 320×440×540 mm