Tisk…
Water stills, mono, Puridest
Dodavatel: LAUDA
L003012 L003014 L003015 L003016 L003018 L003019 L003254 L003245 L003255 L003256 L003258 L003259
171-1303EA 47190 CZK
171-1303 171-1305 171-1306 171-1307 171-1309 171-1310 171-1324 171-1319 171-1325 171-1326 171-1328 171-1329
Water stills, mono, Puridest
Systémy pro výrobu čisté vody Přístroje destilační
Water still suitable for single distillation and for bench mounting.
  • Conductivity stage 1 /25 °C: 2,2 or 2,3 µS/cm
  • Destillate withdrawal through outlet at the condenser
  • Evaporator easily accessible for cleaning purposes
  • CO₂ degassing through outlet in the condenser
  • Thermostatic low water cut-off
  • Manual cooling water control via outlet temperature
  • Evaporator and condenser available in stainless steel or glass
  • Housing made of galvanised sheet steel, powder coated
Economical energy consumption by distillation of the heated cooling water.
Informace proMore Product Information