Tisk…
Percoll™
Dodavatel: Cytiva
17-0891-01 17-0891-02 17-0891-09
17-0891-01EA 16000 CZK
17-0891-01 17-0891-02 17-0891-09
Percoll™
Média k separaci buněk
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose,More Product Information