Tisk…

Sepharose™ CL-4B Octyl Hydrophobic Interaction Chromatography Media

Dodavatel: Cytiva

17-0790-01
17-0790-01EA 23570 CZK
17-0790-01
Sepharose™ CL-4B Octyl Hydrophobic Interaction Chromatography Media
Systémy pro purifikaci proteinů Protein Purification Resins

Octyl-Sepharose™ CL-4B is a well-proven medium for hydrophobic interaction chromatography (HIC). Octyl immobilised on 4% cross-linked agarose.


  • Phenyl immobilized on 4% cross-linked agarose
  • Aromatic ligand
  • Stable in all commonly used aqueous buffers and additives such as 8 M urea and 6 M guanidine hydrochloride
  • Negligible volume variation due to changes in pH or ionic strength


Octyl Sepharose® CL-4B and Phenyl Sepharose® CL-4B are covalently coupled to a cross-linked 4% agarose matrix by ether linkage, resulting in hydrophobic gels with minimal leakage and no ionic properties.

Objednat

Produktová specifikace

Ligand Octyl
Environmentally Preferable
Hustota ligandů 40 μmole/ml drained medium
Matice Cross-linked agarose, 4%, spherical
Průtok 150 cm/h
Rozsah pH 3 - 12
Skladovací teplota 4 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR