Tisk…
Gel filtration media, Sephadex™ G-100
Dodavatel: Cytiva
17-0060-01 17-0061-01
17-0060-01EA 21360 CZK
17-0060-01 17-0061-01
Gel filtration media, Sephadex™ G-100
Systémy pro purifikaci proteinů Protein Purification Resins
Sephadex™ G-100 is useful for desalting and buffer exchange of very large biomolecules. Sephadex™ G-100 Superfine is designed for specific cases requiring very high resolution.
  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Classic gel filtration medium
Sephadex™ is a gel filtration medium prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex™ differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-100 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large biomolecules.
Objednat
Produktová specifikace
Environmentally Preferable
Matice Cross-linked dextran
Maximální tlak 0,096 bar
Rozsah pH 2 - 10