Tisk…
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Dodavatel: Dr. Weigert
410276
148-3112EA 3500 CZK
148-3112
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Prostředky čistící Detergenty a povrchové čističe
neodisher® LaboClean A8.
  • Alkaline powder for universal applications, free of surfactants
  • Components: < 5% chlorine based bleaching agents; 15 to 30% phosphates; 1 to 5% dichloroisocyanuric acid sodium salt
  • Dosage: 2 to 5 g/L
Components: \< 5% chlorine based bleaching agents; 15 - 30% phosphates; 1 - 5% dichloroisocyanuric acid sodium salt.
Dosage: 2 - 5 g/l
Mildly alkaline powder especially suitable for removal of residues of culture media, tissues, blood, proteins and organic dyes. Free fromMore Product Information