Tisk…

Mineral oil, white (petroleum), 75 cSt

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Matrix oil, Mineral oil, Paraffin oil

14621.X2
14621.X2EA 1600 CZK
14621.X2
Mineral oil, white (petroleum), 75 cSt
Mineral oil, white (petroleum)

CAS No.: 8042-47-5 Flash Point: 110°(230°F) Physical Form: Oily liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00131611

Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS číslo: 8042-47-5
EINECS: 232-455-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR