Tisk…

Aluminium oxide, γ-α-phase ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific

14558.14 14558.22 14558.36
14558.14EA 1450 CZK
14558.14 14558.22 14558.36
Aluminium oxide, γ-α-phase ≥99.99% (metals basis)
Aluminium oxide, γ-α-phase

Purity: 99.99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.3 Micron Powder, S.A. 15m2/g Odour: Odourless Merck Reference: 14,356 MDL No.: MFCD00003424

Vzorec: Al₂O₃
Bod varu: 2980 °C (1013 hPa)
Bod tání: 2045 °C
Hustota: 3,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR