Tisk…

Ammonium iodate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

14531.18 14531.30
14531.18EA 2040 CZK
14531.18 14531.30
Ammonium iodate ≥99%
Ammonium iodate

Purity: 99% CAS No.: 13446-09-8 Molecular Formula: H4INO3 Formula Weight: 192.94 Physical Form: Crystalline Density: 3.309 Appearance: White Odour: Odourless Melting Point: 150° UN No.: 1479 MDL No.: MFCD00049684

Vzorec: H₄INO₃
Bod tání: 150 °C
Hustota: 3,309
Storage Temperature: Ambient
MDL Number: MFCD00049684
CAS číslo: 13446-09-8
EINECS: 236-592-4
UN: 1479
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR