Tisk…

Oxid zinečnatý 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Dodavatel: Honeywell Chemicals

Synonyms: Zinci oxidum, Zinc (II) oxide

14439-1KG
14439-1KGEA 1840 CZK
14439-1KG
Oxid zinečnatý 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zinc oxide
Vzorec: ZnO
Hustota: 5,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS číslo: 1314-13-2
EINECS: 215-222-5
ADR: 9,III

Objednat

Výsledky specifikačního testu

Assay (calculated to ignited substance) 99.0 - 100.5 %
Loss on ignition (500°C) Max. 1.0 %
Alkalinely reacting substances complying
Arsenic (As) Max. 3 ppm
Cadmium (Cd) Max. 10 ppm
Iron (Fe) Max. 10 ppm
Lead (Pb) Max. 50 ppm
Lead complying
Iron and other heavy metals complying
Carbonate and color of the solution complying
Carbonates and substances insoluble in acids complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR