Tisk…

Manganese(II) carbonate ≥99.9% (metals basis excluding Na)

Dodavatel: Thermo Fisher Scientific

14344.30 14344.A1
14344.30EA 2000 CZK
14344.30 14344.A1
Manganese(II) carbonate ≥99.9% (metals basis excluding Na)
Manganese(II) carbonate

Purity: 99.9% CAS No.: 598-62-9 Molecular Formula: MnCO3 Formula Weight: 114.95 Physical Form: Powder Density: 3.125 Appearance: Light brown to violet Odour: Odourless Melting Point: 350° dec. Merck Reference: 14,5726 MDL No.: MFCD00011116

Vzorec: CMnO₃
Hustota: 3,125
Storage Temperature: Ambient
MDL Number: MFCD00011116
CAS číslo: 598-62-9
EINECS: 209-942-9
Merck Index: 14,5726

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR