Tisk…

Magnesium ethanolate (21 - 22% Mg)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

14264.30 14264.A1
14264.30EA 4360 CZK
14264.30 14264.A1
Magnesium ethanolate (21 - 22% Mg)
Magnesium ethanolate

CAS No.: 2414-98-4 Molecular Formula: C4H10MgO2 Formula Weight: 114.44 Physical Form: Granular Density: 1.01 Sensitivity: Air and Moisture Sensitive Appearance: Off-white Odour: Odourless Melting Point: 270° dec. Beilstein Reference: 3678841 UN No.: 3205 MDL No.: MFCD00009060

Vzorec: C₄H₁₀MgO₂
Hustota: 1,01
Storage Temperature: Ambient
MDL Number: MFCD00009060
CAS číslo: 2414-98-4
EINECS: 219-323-5
UN: 3205
ADR: 4.2,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR