Tisk…

Potassium ethanolate ≥95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Potassium ethylate, Potassium ethoxide

14263.30 14263.A1
14263.30EA 4600 CZK
14263.30 14263.A1
Potassium ethanolate ≥95%
Potassium ethanolate

Purity: 95% CAS No.: 917-58-8 Molecular Formula: C2H5KO Formula Weight: 84.16 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow to pale brown Odour: Alcohol-like Melting Point: 250° dec. Solubility: Soluble in ethanol, ether UN No.: 3206 MDL No.: MFCD00054277

Vzorec: C₂H₅KO
Storage Temperature: Ambient
MDL Number: MFCD00054277
CAS číslo: 917-58-8
EINECS: 213-029-0
UN: 3206
ADR: 4.2,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR