Tisk…

Boron standard dissolved in oil, 1.000 µg/g B in mineral oil Specpure® pro wear metal analysis

Dodavatel: Thermo Fisher Scientific

14227.18
14227.18EA 3470 CZK
14227.18
Boron standard dissolved in oil, 1.000 µg/g B in mineral oil Specpure® pro wear metal analysis
Standardy Standardy kationické Standrady - bor

Physical Form: Liquid
MDL No.: MFCD00134034

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR