Tisk…

Cleaning agent, TICKOPUR RW 77

Dodavatel: BANDELIN-ELECTRONIC
898
142-1953EA 1350 CZK
142-1953
Cleaning agent, TICKOPUR RW 77
Prostředky čistící Detergenty a povrchové čističe
Cleaning agent suitable for cleaning of ultrasonic bath. It can be used to clean glass, metal, ceramics, plastic, rubber and removes grease, oils, wax, protein, pigments, etc.

  • Contains ammonia
  • pH 9,9 at 1%
  • Emulsifying
  • Phosphate-free
Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR