Tisk…
Ultrasonic baths, Bransonic®
Dodavatel: BRANSON
CPX-952-336R CPX-952-837R CPX-952-836R CPX-952-537R CPX-952-536R CPX-952-136R CPX-952-337R CPX-952-237R CPX-952-236R CPX-952-137R
142-0133EA 36460 CZK
142-0133 142-0140 142-0139 142-0137 142-0136 142-0127 142-0134 142-0131 142-0130 142-0128
Ultrasonic baths, Bransonic®
Lázně ultrazvukové
M and MH Series ultrasonic baths are known for offering a high level of precision and reliability.
  • Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank
  • 40 kHz rugged industrial transducers
  • Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions
  • Plastic body, chemical-resistant
M Series includes 60-min mechanical timer and continuous operation.
MH Series includes 60-min mechanical timer andMore Product Information