Tisk…
Brushes, FlexBrush™
Dodavatel: MICRONOVA
FB-12 FBC-611 FBC-611IR BRWM-22512J BRWM-22512S_U FBC-6AA
129-1068EA 1660 CZK
129-1068 141-1016 141-1644 129-4202 MICRBRWM-22512S_U 141-1645
Brushes, FlexBrush™
Kartáče
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Objednat
Produktová specifikace
Environmentally Preferable Waste Reducing