Tisk…
Cleaning trolley Clino®, Profi C9
Dodavatel: PFENNIG REINIGUNGSTECHNIK
2099970
141-1389EA 48190 CZK
141-1389
Cleaning trolley Clino®, Profi C9
Vozíky
The Clino® Profi C9 ergonomically optimised system trolley is exclusively developed and used for EasyMop-technique. It is made of stainless steel.
  • Floor cleaning is simple with flat wringer Ringo and mop-reuse-technique
  • Lifetime durability through stainless stell tubular frame construction
  • Disposal unit at rest can be folded in, every time exchangeable and expandable
  • Highest efficiency and process reliability in single-step technique
  • Single-stage, used for one wiping process
  • Double ball-bearinged guide wheels (125 mm)
Informace o dodávce: SuppliedMore Product Information