Tisk…
Cleanroom vacuum cleaner, MV-1CR (CV)
Dodavatel: TIGER-VAC
110744B 110745B 110748B
141-1368EA 84260 CZK
141-1368 141-1369 141-1370
Cleanroom vacuum cleaner, MV-1CR (CV)
Vysavače
Designed for general clean-up in contamination controlled environments, cleanroom compatible and ideal for the pharmaceutical industry. Available in Handheld (HH), Backpack (BP) and Golfcart (GC) versions.
  • Single phase thru-flow motor
  • Recovery capacity: 0,4-1,2 litres within the containment HEPA filter
This unit is equipped with a true ULPA filter (U15) located upstream of the motor. It also contains a containment HEPA filter (H14). The HEPA filter is sealed directly to the lid of the vacuum to ensure entirely leak-free operation.
It is EMI/RFIMore Product Information